Wika Non-Incendive Pressure Transmitters
This is an FM approved non-incendive for Class I, Division II, Groups A, B, C, and D. It features a 1/2''male conduit electrical connection, NACE listed wetted parts, NEMA 4 / IP 67 weather protection, and excellent vibration resistance.
$491.93USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Process Connection
Availability Pricing
Wika N-10 Non-Incendive Pressure Transmitter, 0 to 30 psig EW-68446-43 Mfr # 4348326
COMPARE
±0.25% full-scale (BFSL) 30 1/4\" NPT(M)
$491.93USD / Each