Winters Tube Stub Gauges
Use gauges with standard double ferrule tube fittings
Heavy-duty, stainless steel, liquid-filled gauges are designed for the laboratory where a standard NPT fitting cannot be used, such as when a Parker® A-Lok® is the standard process connection or where a PTFE-free connection is desired. The tube stub fittings eliminate leaking at the connection point. Place the gauge in any orientation, even where space is limited.
Other pressure ranges and mounting configurations are available. Contact an Application Specialist for more information.
$123.25USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
Compareitem specifications
Accuracy Max Pressure (PSI) Resolution
Availability Pricing
Winters Instruments PTS820-TS Tube Stub Gauge, Stainless Steel, 2-1/2" Dial, Vacuum EW-68076-22 Mfr # PTS820-TS
COMPARE
±1.5% full-scale
$123.25USD / Each