Work2Store® Expandable Tube Racks
No more rack clutter or cross-contamination
  • Sliding hinges expand storage rack into working rack
  • Compact storage configuration fits in 2" freezer racks
  • Wide spaces between rows in working configuration reduces interference and errors
  • Storage configuration dimensions: 5-1/4"W x 2-1/16"H x 5-1/4"D (13.3 x 5.3 x 13.3 cm)
  • Working configuration dimensions: 12-3/16"W x 1-5/16"H x 5-1/16"D (31 x 3.4 x 12.8 cm)
These racks function as both storage and working tube holders, so you don't need extra racks taking up valuable space. Samples stay in one place, minimizing cross-contamination common when moving tubes from one rack to another. When hinges are collapsed, the rack fits standard 2" freezer racks, no special racking system required. When hinges are expanded, spacing between wells is wide enough to accommodate open hinged-lid tubes without obscuring tubes in the next row. Extra space between rows also makes it easier to differentiate between tubes, reducing errors in transferring and titrating.
Additional features include a locking lid to secure the rack in the storage configuration, a center depression in each well to stabilize conical-bottom tubes, and a cryogenic vial locking mechanism to enable one-handed cap removal.
$42.30USD / Each
1 variations of this product are available.
See All Product Options Below
1 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Tubes Held Description
Availability Pricing
Heathrow Scientific Work2Store™ Working and Storage Rack, 64 x 1.5 / 2.0 mL cryo tubes, 1/ea EW-06696-11 Mfr # HS120018
COMPARE
64 Working and Storage Rack, 64 x 1.5 / 2.0 mL cryo tubes, 1/ea
$42.30USD / Each