Xtra-Value Electronic Caliper Gauges
Displays measurements in inches, millimeters, or fractions
  • Spring-loaded travel jaws provide smooth operation and movement
  • Auto off feature maximizes battery life
  • Available for internal or external caliper gauge versions
  • Resolution: 0.01", 0.3 mm, or 1/64
Robust housing holds the chrome-finished alloy jaws in place to obtain accurate and reliable internal or external measurements. Easily find the thinnest or widest dimension and freeze the reading in place with the thumb-screw lock nut, and or by pressing Hold on the gauge. With the press of a button, the large LCD converts inches to millimeters — or even fractions.
$71.60 - $77.50USD / Each
2 variations of this product are available.
See All Product Options Below
2 PRODUCT OPTIONS
  • filter by:
Compareitem specifications
Max range (in) Resolution
Availability Pricing
Fowler 54-554-730 Xtra-Value Electronic Caliper Gauge, 6 in Length; Internal EW-97604-83 Mfr # 545547300
COMPARE
6 0.01 in or 1/64 in
$77.50USD / Each
Fowler 54-554-630 Xtra-Value Electronic Caliper Gauge, 6 in Length; External EW-97604-84 Mfr # 545546300
COMPARE
6 0.01 in or 1/64 in
$71.60USD / Each