Cole-Parmer® Remote-Mount Control Flowmeters
2/20/2007
Perform proportional feed and batch processing accurately and easily with the new Cole-Parmer remote-mount control flowmeters. Newly introduced and featuring an analog output, these meters are ideal for water/wastewater treatment, chemical systems, and agricultural applications with water-like fluids containing less than 1% solids. To suit a wide range of pipe sizes, meters are offered with in-line polypropylene or saddle-mount PVDF pipe fittings. Eight-digit LCD can be mounted directly to the pipe or into a ¼-DIN panel opening.