Cole-Parmer® Wide-View™ Analog Panel Meters
7/20/2006
Designed specifically for temperature and pH measurement
Cole-Parmer introduces a new line of analog panel meters designed for temperature and pH measurement in a variety of ranges. These high-quality meters are a great alternative to higher cost digital meters and are perfect for any temperature or electrochemical measurement application. Panel meter models 65550-00 to -88 feature a molded black phenolic case and clear acrylic window that provides wide-angle visibility. All units have ±2% full scale accuracy and meet ANSI specification C39.1. Panel meters accept inputs of 4-to-20 mA, 1-to-5 VDC, or 10-to-30 VDC.