Davis Instruments presents Advanced Modular Calibration Systems from GE Sensing®
11/3/2008
Powerful and flexible, new GE Druck DPI 620 Multifunction Calibrators provide world-class measurement and generation
Davis Instruments is proud to introduce new DPI 620 Multifunction Calibrators from GE Sensing. These GE Sensing advanced modular calibration systems provide power and precision in a robust, portable design.
The DPI 620 Multifunction Calibrators bring flexibility to the fold, able to simultaneously measure and source mA, mV, V, ohms, frequency, and a variety of RTDs and thermocouples. Built-in HART® configurator provides setup and calibration adjustment of HART devices for digital communication. Each calibrator is capable of measuring pressures from 0.35 to 15,000 psi.
Easy-to-use video-quality touch screen provides a bright, crystal-clear display which can be operated by a gloved hand. Built with strong casing, weatherproof to IP65, these calibrators are able to withstand tough conditions on the production floor or out in the field.
Find these multifunction calibrators and over 50 other GE Sensing products on Davis.com, including pressure stations, pressure modules, and accessories. Davis Instruments has many GE Sensing items in stock and ready to ship, with next-day shipping available.
To see Davis Instruments’ entire range of GE Sensing products—as well as a complete selection of industry-leading test, measurement, control, and calibration instruments—get the new Davis Instruments V74 Sourcebook. To request your FREE copy, call 800-358-5525 or visit Davis.com/3953