New Complete-System Drum Pumps
7/12/2004
Pump tube and motor as one unit
Cole-Parmer Instrument Company introduces a new line of Cole-Parmerr drum and container pump systems that come with pump tube and motor sold as one unit. These pump systems are equipped with variable-speed control motors, and have no mechanical seal within the pump tube-reducing overall maintenance.
Order polypropylene, PVDF, CPVC, or 316 SS pump tubes in lengths of 39" or 47". Optional components include barrel adapter, strainer, wall mounting bracket, and dispensing nozzle with six feet of hose.
Download 300 DPI 7061340.jpg Image