Cole-Parmer Instrument Company introduces the new Digi-Sense infrared thermometer with type K thermocouple input and Class II laser sighting for measuring and displaying non-contact and contact temperatures simultaneously.
Unit features a distance-to-target ratio of 11:1, with an adjustable emissivity range of 0.1 to 1 for a wider selection of surface measurements. The backlit LCD simultaneously displays the current temperature in
Measures temperatures as high as 1999
Download 300 DPI 3964400.jpg Image