New Oakton® Basic CON 11 and Advanced CON 110 Conductivity and Temperature Meters
5/16/2004
Units offer increased measurement and temperature ranges, plus more data storage
Cole-Parmer Instrument Company introduces the new CON 11 and CON 110 conductivity and temperature meters, the latest upgrades to our popular Oakton line of handheld meters.
Both meters offer user-friendly operation and feature large, dual-line LCDs that display the measured parameter, temperature, graphic symbols, and self-diagnostic message codes. In addition, these meters offer auto-range functions that select the proper measurement range with ±1% full-scale accuracy. Built-in stand and optional AC/DC adapter allow for laboratory or field use.
Improvements to the CON 11 meter include single- or multi-point automatic and manual calibration, increased conductivity range of 0 to 199.9 mS/cm, increased TDS range of 0 to 99.9 ppt, and increased temperature range of -10 to 110?C. In addition, the meter now has programmable TDS factors of between 0.4 and 1.0, and it accepts conductivity electrodes with cell constants of 0.1, 1.0, or 10.0. The unit also has increased battery life, and increased data storage, allowing up to 50 data sets for recall.
The CON 110 has all the improvements mentioned above, plus it measures temperature in ?C or ?F, has an RS-232 output, and comes with complimentary data acquisition software for easy downloads. The CON 110 also stores 100 data sets for recall or for transfer to a printer or computer for analysis.
Download 300 DPI 3560745.jpg Image