Lower-priced alternative to stainless steel model—with all the functionality
Vernon Hills, Ill., July 2, 2013—Introducing the Masterflex® L/S® Digital Console Process Drives from Cole-Parmer, housed in powder-coat steel for a more economical price. These peristaltic pumps are ideal for fluid transfer, dispensing, and metering applications in laboratory, process, and washdown environments. The housings are completely sealed and IP66 and NEMA 4X rated.
These precision tubing pumps offer analog remote control options and simple programming—ideal for automated process applications. The pumps' brushless, maintenance-free motor has ±0.1% speed control accuracy with a 6000 to 1 turndown ratio. It features a programmable dispense interval that lets users set the delay between dispense cycles for convenient, automated dispensing. Programmed calibration ensures dispense and display accuracy—the drive stores one calibration value per tubing size. The motor is reversible to purge tubing before or after pumping and to pump fluid in either direction.
The graphical LCD shows four operating modes: continuous run, timed dispense, copy dispense, volume dispense. Users can view pump performance continually. The drive is programmable via the simple membrane keypad with keypad lock/unlock feature to protect settings. Its menu includes seven languages for programming and operation.
The easy-to-clean washdown drives are resistant to common cleaning and sanitizing solutions including chlorine-based sanitizers. Simply hose down for easy maintenance. For more information on the Masterflex® L/S® Digital Console Process Drives from Cole-Parmer, call 800-323-4340 or go to ColeParmer.com/20159.
Cole-Parmer has been a leading global source of laboratory and industrial fluid handling products, instrumentation, equipment, and supplies since 1955. Our product lines, including popular brand names such as Masterflex®, Oakton®, and more, are sold through company-owned customer channel outlets and a strong network of international dealers. We also feature an ISO-17025-accredited metrology lab for instrument calibration and repair. Cole-Parmer responds with excellence to customer needs, and offers application expertise and technical support. For more information, contact Cole-Parmer, 625 East Bunker Court, Vernon Hills, IL 60061. In the US, call 800-323-4340. International customers, call 847-549-7600. Visit us at ColeParmer.com
Download 300 DPI 07575_20_el2head.jpg Image