Products Site Map 2

Labchem Hydroxylamine Hydrochloride 10 W V
Labchem Hydroxylamine Hydrochloride 50 W V
Labchem Hydroxynaphthol Blue
Labchem Iron Aa Standard
Labchem Isopropyl Alcohol 50 V V
Labchem Lactic Acid
Labchem Lead Aa Standard
Labchem Lithium Aa Standard
Labchem Magnesium Aa Standard
Labchem Manganese Aa Standard
Labchem Manganous Sulfate Solution For Dissolved Oxygen
Labchem Mercuric Nitrate 0 141 N 0 071 M For Chloride
Labchem Mercuric Nitrate 0 1 N 0 05 M
Labchem Mercuric Thiocyanate 0 42 W V In Methanol
Labchem Mercury Aa Standard
Labchem Methanol 50 V V 1 1
Labchem Methylene Blue 0 1 Aqueous
Labchem Methylene Blue 1 In Ethanol
Labchem Methylene Blue Reagent For Surfactants
Labchem Methyl Red 0 02 Aqueous
Labchem Methyl Red 0 02 In Methanol
Labchem Methyl Red 0 1 Aqueous
Labchem Methyl Red 1 0 Aqueous
Labchem Methyl Red Methylene Blue Mixed Indicator For Nitrogen Ammonia Organic
Labchem Molybdenum Aa Standard
Labchem New Methylene Blue N For Reticulocytes
Labchem Nickel Aa Standard
Labchem Nitrate As Nitrogen Standard
Labchem Nitric Acid 0 1 N 0 1 M
Labchem Nitric Acid 1 0 N 1 0 M
Labchem Nitric Acid 10 V V 1 9
Labchem Nitric Acid 50 V V 1 1
Labchem Nitrite As Nitrogen Standard Preserved With Chloroform
Labchem O Tolidine 0 1
Labchem Oxalic Acid 10 W V For Silica
Labchem Oxalic Acid 5 W V For Silica
Labchem Oxalic Acid 7 5 W V For Silica
Labchem Phenol 5 W V
Labchem Phenolphthalein 0 08 In Methanol
Labchem Phosphate As Phosphorus Standard
Labchem Phosphate Buffer Ph 4 1 For Cyanide
Labchem Phosphate Buffer Ph 6 2 6 5 For Chlorine Chlorine Dioxide
Labchem Phosphate Buffer Ph 6 2 6 5 Mercury Free For Chlorine Chlorine Dioxide
Labchem Phosphate Buffer Ph 7 0 For Chlorine Chlorine Dioxide
Labchem Phosphate Standard
Labchem Phosphoric Acid 25 V V 1 3
Labchem Potassium Aa Standard
Labchem Potassium Antimony Tartrate Solution For Phosphorus
Labchem Potassium Biiodate 0 025 N 0 0021 M
Labchem Potassium Biiodate 0 0375 N 0 00313 M
Labchem Potassium Biiodate 0 1 N 0 0085 M
Labchem Potassium Chromate Indicator 5 W V For Chloride
Labchem Potassium Dichromate 0 25 N 0 0417 M
Labchem Potassium Ferricyanide 10 W V
Labchem Potassium Ferrocyanide 10 W V
Labchem Potassium Hydroxide
Labchem Potassium Hydroxide 0 01 N 0 01 M
Labchem Potassium Hydroxide 0 1 N 0 1 M
Labchem Potassium Hydroxide 0 1 N 0 1 M In Ethanol
Labchem Potassium Hydroxide 0 5 N 0 5 M
Labchem Potassium Hydroxide 0 5 N 0 5 M In Isopropanol
Labchem Potassium Hydroxide 0 5 N 0 5 M In Methanol
Labchem Potassium Hydroxide 1 0 N 1 0 M
Labchem Potassium Hydroxide 10 N 10 M
Labchem Potassium Hydroxide 1 0 N 1 0 M In Methanol
Labchem Potassium Hydroxide 10 W V
Labchem Potassium Hydroxide 2 0 N 2 0 M
Labchem Potassium Hydroxide 20 W V
Labchem Potassium Hydroxide 30 W V
Labchem Potassium Hydroxide 45 W W
Labchem Potassium Hydroxide 50 W V
Labchem Potassium Iodate 0 1 N 0 0167 M
Labchem Potassium Iodide
Labchem Potassium Iodide 0 5 W V For Chlorine Chlorine Dioxide
Labchem Potassium Iodide 15 W V For Arsenic
Labchem Potassium Iodide 20 W V
Labchem Potassium Permanganate 0 588 N 0 1176 M
Labchem Potassium Permanganate 0 5 N 0 1 M
Labchem Potassium Permanganate 4 W V For Chromium
Labchem Potassium Persulfate 5 W V For Mercury Metals
Labchem Potassium Thiocyanate 0 1 N 0 1 M
Labchem Potassium Thiocyanate 10 W V
Labchem Pyridine
Labchem Safranin 1 W V Aqueous
Labchem Selenium Aa Standard
Labchem Silica Standard
Labchem Silver Nitrate
Labchem Silver Nitrate 0 0171 N 0 0171 M
Labchem Silver Nitrate 0 0192 N 0 0192 M For Cyanide
Labchem Silver Nitrate 0 01 N 0 01 M
Labchem Silver Nitrate 0 141 N 0 141 M
Labchem Silver Nitrate 0 153 N 0 153 M
Labchem Silver Nitrate 0 1 N 0 1 M
Labchem Silver Nitrate 0 5 N 0 5 M
Labchem Silver Nitrate 1 0 N 1 0 M
Labchem Sodium Aa Standard
Labchem Sodium Acetate 2 0 M 2 0 N For Zinc
Labchem Sodium Acetate 20 W W For Iron
Labchem Sodium Arsenite 0 1 N 0 05 M For Residual Chlorine
Labchem Sodium Azide 0 5 W V For Chromium
Labchem Sodium Carbonate 1 0 N 0 5 M
Labchem Sodium Carbonate 10 W V
Labchem Sodium Chloride 0 1 N 0 1 M
Labchem Sodium Chloride 0 85 W V
Labchem Sodium Chloride 20 W V For Iodide
Labchem Sodium Chloride 25 W V
Labchem Sodium Chloride 30 W V
Labchem Sodium Chloride Hydroxylamine Sulfate For Metals
Labchem Sodium Hydroxide
Labchem Sodium Hydroxide 0 01 N 0 01 M
Labchem Sodium Hydroxide 0 0227 N 0 0227 M
Labchem Sodium Hydroxide 0 05 N 0 05 M
Labchem Sodium Hydroxide 0 25 N 0 25 M
Labchem Sodium Hydroxide 0 3125 N 0 3125 M
Labchem Sodium Hydroxide 0 333 N 0 333 M
Labchem Sodium Hydroxide 10 0 N 10 0 M
Labchem Sodium Hydroxide 10 W V
Labchem Sodium Hydroxide 10 W W
Labchem Sodium Hydroxide 3 0 N 3 0 M
Labchem Sodium Hydroxide 33 W W
Labchem Sodium Hydroxide 4 0 N 4 0 M
Labchem Sodium Hydroxide 5 0 N 5 0 M
Labchem Sodium Hydroxide 50 W V
Labchem Sodium Hydroxide 5 W V
Labchem Sodium Hypochlorite 2 3 W V
Labchem Sodium Nitroprusside 0 05 W V For Nitrogen
Labchem Sodium Phenate Solution For Nitrogen Ammonia Organic
Labchem Sodium Sulfate Anhydrous
Labchem Sodium Thiocyanate 0 1 N 0 1 M
Labchem Sodium Thiosulfate 0 01 N 0 01 M
Labchem Sodium Thiosulfate 0 05 N 0 05 M
Labchem Sodium Thiosulfate 1 0 N 1 0 M
Labchem Sodium Thiosulfate 2 0 N 2 0 M
Labchem Stannous Chloride 40 For Arsenic
Labchem Starch Indicator 0 5 Preserved With Chloroform
Labchem Starch Indicator 1 Preserved With Chloroform
Labchem Strontium Aa Standard
Labchem Sulfamic Acid
Labchem Sulfanilamide 1 W V For Nitrogen Nitrate Nitrite
Labchem Sulfanilamide N Naphthyl Reagent For Nitrogen Nitrate Nitrite
Labchem Sulfuric Acid 0 04 N 0 02 M
Labchem Sulfuric Acid 0 05 N 0 025 M
Labchem Sulfuric Acid 0 1 N 0 05 M
Labchem Sulfuric Acid 0 255 N 0 1275 M
Labchem Sulfuric Acid 1 0 N 0 5 M
Labchem Sulfuric Acid 10 V V 1 9
Labchem Tclp Extraction Fluid 1
Labchem Tclp Extraction Fluid 2
Labchem Thymolphthalein 0 05 In Ethanol
Labchem Tin Aa Standard
Labchem Toluidine Blue 1 Aqueous
Labchem Trichloroacetic Acid 10 W V
Labchem Turbidity Stock Solution I Hydrazine Sulfate 1
Labchem Turbidity Stock Solution Ii Hexamethylene Tetramine 10
Labchem Vanadium Aa Standard
Labchem Vanadomolybdate Solution In Dilute Perchloric Acid
Labchem Xylenes
Lactic Acid Lithium Salt
Lanthanum Iii Nitrate Hexahydrate
Lanthanum Oxide
L Arginine
L Arginine Hydrochloride
L Ascorbic Acid
Lauroyl Chloride
Lb Agar Miller
Lb Broth
Lb Broth Lennox
L Borneol
L Dopa
Lead Ii Acetate Trihydrate
Lead Ii Bromide
Lead Ii Iodide
Lead Ii Nitrate
Lever Drum Pumps
L Glutamic Acid Monosodium Salt Monohydrate
L Glutamine
L Homoarginine Hydrochloride
Linktech 20Ac 20Pp Series Quick Disconnect Fitting Body Hose Barb Without Valve
Linktech 20Ac And 20Pp Series Quick Disconnect Fitting Insert Hose Barb Without Valve Panel Mount
Linktech 20Cb Series Quick Disconnect Fitting Body Npt M With Valve
Linktech 20Cb Series Quick Disconnect Fitting Insert Bspt M With Valve
Linktech 20Cb Series Quick Disconnect Fitting Insert Npt F Without Valve
Linktech 30Ac Series Quick Disconnect Fittings Body Hose Barb With Valve Panel Mount
Linktech 30Ac Series Quick Disconnect Fittings Insert Npt M Without Valve
Linktech 40Ac And 40Pp Series Quick Disconnect Fitting Insert Hose Barb Without Valve Elbow 68090
Linktech 40Ac And 40Pp Series Quick Disconnect Fitting Insert In Line Hose Barb With Valve
Linktech 40Ac And 40Pp Series Quick Disconnect Fitting Insert O D Tubing With Valve Elbow
Linktech 40Cb Series Quick Disconnect Fittings Body Hose Barb With Valve
Linktech 40Cb Series Quick Disconnect Fittings Insert Hose Barb Without Valve Elbow
Linktech 50Ac Series Quick Disconnect Fitting Body Compression With Valve
Linktech 50Ac Series Quick Disconnect Fitting Insert Compression With Valve 90 Elbow
Linktech 60Pp And 60Ps Series Quick Disconnect Fitting Body Npt M With Valve 68296
Lithium Acetate Dihydrate
Lithium Plasma Grade Reference Standard
Lithium Stearate
Lithium Tetrafluoroborate
Lutetium
Madgetech Process101A Current Data Loggers
Magnesium Oxide 23299
Magnesium Plasma Grade Reference Standard 39587
Main Body Assemblies 4653
Make A Clamp Kit
Maleic Acid 26096
Manganese Standard
Manganous Sulfate 0 00300 Molar
Mass Flow Command Modules
Masterflex L S High Performance Precision Biopharm Silicone Platinum Cured Pump Tubing
Masterflex L S High Performance Precision Pharmed Bpt Pump Tubing For Masterflex L S Pumps
Masterflex L S High Performance Precision Tygon Chemical 2001 Pump Tubing
Masterflex L S Precision Biopharm Plus Silicone Platinum Cured Pump Tubing
Masterflex L S Precision Norprene A 60 G Pump Tubing For Masterflex L S Pumps
Masterflex L S Precision Platinum Cured Silicone Pump Tubing
Masterflex L S Precision Silicone Platinum Cured Pump Tubing For Masterflex L S Pumps
Masterflex L S Precision Tygon E Lab E 3603 Pump Tubing For Masterflex Pumps
Masterflex L S Precision Viton Fda Compliant Pump Tubing For Masterflex L S Pumps
Masterflex L S Precision Washdown Modular Drives With Wall Mount Controller
Masterflex Plastic Concentric Sanitary Clamp Reducers
Masterflex Straight Barbed Reducing Connectors
Mbth Indicator 0 05 W V Aqueous Solution
M Cresol
Mecmesin Rs 232 Communication Cable
Mercaptoacetic Acid
Mercaptoacetic Acid Sodium Salt
Mercuric Nitrate 0 100 Normal N 10
Meso 2 3 Dibromosuccinic Acid
Metal Quick Disconnect Fittings Hose Barb Elbow Inserts
Methyl 4 Hydroxybenzoate
Methylacrylate
Methyl Alpha D Mannopyranoside
Methylene Blue 1 W V Aqueous Solution 39640
Methyl Formate
Methyl Lactate
Methyl Methanesulfonate
Methyl Myristate
Methyl Red Hydrochloride
Methylthymol Blue Tetrasodium Salt
Methyltriethoxysilane
Methyltrimethoxysilane
Mettler Toledo Accessories For Densito Series Density Meters
Mettler Toledo Benchtop Digital Refractometer
Mettler Toledo S230 Sevencompact Benchtop Conductivity Meters
Microflex Comfortgrip Latex Gloves
Microflex Diamond Grip Plus Latex Gloves
Microflex Freeform Se Nitrile Gloves
Microflex Synetron Latex Gloves
Microflex Ultra One Latex Gloves
Micropump Gear Pump Skid Systems With Calibration Column
Mid West Instruments Gauge Minder Pressure Limiting Valve Dampener
Mid West Model 142 Diaphragm Differential Pressure Gauge And Transmitter
Mineral Oil Heavy
Mobile Computer Stand
Monarch Instrument Electronic Paperless Recorders
Morpholine
Mounting Kits For Industrial Ethernet Switches
M System 53U Multi Lcd Power Monitor
M System Limit Alarms
Multifunction Portable Calibrators
Multipro Sector Probes
Myristic Acid
Myronl Ph Ultrapen Testers
Nabertherm Muffle Furnaces
N Acetyl L Tyrosine Ethyl Ester Monohydrate
Nalidixic Acid
Naphthenic Acids 37136
N Bromosuccinimide 36893
N Butylamine
Neodymium Atomic Absorption Standard Solution
Neodymium Standard
Neomycin Sulfate 26139
N Hexadecane
Nickel Ii Chloride Hexahydrate 23354
Nickel Standard
Nicotinic Acid
Nicotinic Acid N Oxide
Nitrilotriacetic Acid 26147
N Methyl N Trimethylsilyl Trifluoroacetamide
N N Diethylethanolamine
N N Dimethyl 3 Nitroaniline
N N Diphenylbenzidine
Norit R Sa Ii
Notice Turn Off Cellular Phone Pagers And Other Wireless Devies Signs
N Pentadecanoic Acid
N Propyl Acetate 37107
N Tron 508 Series Industrial Ethernet Switches
N Tron Industrial Ethernet Switch 59714
N Tron Industrial Ethernet Switches With Poe
N Tron Industrial Poe Mid Span Injector
N Tron Industrial Wireless Ethernet Device
N Tron Managed Gigabit Capable Industrial Ethernet Switches
Oakton Electrode Cleaning Solutions
O Cresol 23362
Octadecylbenzene
Ohaus Adventurer Ax Ntep Approved Toploading Balances
Ohaus Anti Theft Devices
Ohaus Auxiliary Displays
Ohaus Aviator 7000 Price Computing Scale
Ohaus Breeze Break Draft Shields
Ohaus Ranger 4000 Compact Bench Scale
Ohaus Valor 3000 Portion Control Scales
Ohaus Valor 3000 Portion Control Scales Ntep
Oleic Acid Sodium Salt
Optical Tachometer Reflective Tape
Opto 22 Snap Brains
Orcinol
O Toluidine
Oxalyl Chloride
Oxygen And Toxic Gases Monitor Alarm
Pacific Transducer Durometer Test Stand
Palladium Ii Acetate
Palladium On Alumina Powder
Panel Mount Compression Inserts 57171
Panel Mount Hose Barb Bodies With 1 4 Flow
Partlow Recorders Accessories
Pelican L4 Led Flashlights
Penicillin G Potassium Salt
Pentafluoropropionic Acid
Pentamethylbenzaldehyde
Peroxide Cured Silicone Tubing Blue
Pfa Sanitary Clamp Reducers
Pfa Sanitary Clamp Tees
Phase Ii Digital Coating Thickness Gauges
Phenethyl Alcohol
Phenol Red Indicator 0 04 W V Aqueous Solution
Phenol Saturated
Phenol Solutions
Phenoxyacetic Acid
Phenylarsine Oxide Pao Titrant
Phenyl Benzoate 37558
Phenylphosphonic Acid
Phloroglucinol Dihydrate
Phosphorous Acid
Phosphorus Oxychloride
Phosphorus Pentoxide
Phosphorus Standard
Phosphotungstic Acid
Phosphotungstic Acid 44 Hydrate
Pinacolone 37660
Plastic Quick Disconnect Fittings Sealing Plug
Platinum Cured Silicone Tubing Spools
Plug Caps 4159
Polarization Dissolved Oxygen Sensors
Polarsafe Eva Ice Pans
Poly Acrylamide
Poly Caprolactone Diol
Polyfoil Square Wrap
Poly Methyl Methacrylate 37733
Polypropylene Adapter Heads 18362
Polyscience Coliform Testing Circulating Baths
Polyscience Heated Circulating Baths
Poly Sodium P Styrenesulfonate
Polystyrene Slide Holders
Polysulfone Holder
Polyurethane Stools
Poly Vinyl Alcohol
Potassium Acid Phthalate 0 0500 Normal N 20
Potassium Acid Phthalate 300 Mg L Multi Purpose Standard
Potassium Bisulfate
Potassium Dichromate 0 0100 Normal N 100
Potassium Hexachloroplatinate Iv
Potassium Hydroxide 0 100 Normal N 10
Potassium Hydroxide 0 100 Normal N 10 In Isopropyl Alcohol
Potassium Hydroxide 0 100 Normal N 10 In Methanol
Potassium Hydroxide 0 200 Normal N 5
Potassium Hydroxide 0 250 Normal N 4
Potassium Hydroxide 0 500 Normal N 2 In Methanol
Potassium Hydroxide 1 00 Normal
Potassium Hydroxide 1 00 Normal In Denatured Alcohol
Potassium Hydroxide 10 W V Aqueous Solution 39818
Potassium Permanganate 1 00 Normal
Potassium Permanganate 47241
Potassium Permanganate 4 W V Aqueous Solution
Potassium Permanganate Solution 23417
Potassium Plasma Grade Reference Standard
Potassium Tetrachloropalladate Ii
Pressure Calibrators Single Or Dual Range
Pressurements Hydraulic Deadweight Testers
Primelock High Purity Pfa Fittings Spacesaver Elbows
Progesterone
Propargyl Alcohol
Propylene Carbonate
Propylene Oxide
Protractors
Prp 1 Reverse Phase Columns
P Terphenyl 37401
Ptfe Straight Nptf Couplings
Pulsafeeder Service Kits
Pump And Mix Tube Sets
Pyrazinamide
Pyrex Brand 6180 Funnels
Pyridine N Oxide
Pyrocatechol Violet 37891
Pyruvic Acid
Pyruvic Acid Sodium Salt
Qorpak Kaptclean Sample Jars
Quality Process Gauge
Quercetin Dihydrate
Quinine
Rack Support System Accessory Screws
Rae Toxirae Pro Single Gas Detector
Red Lion Epax Extra Large Digit Panel Meters
Red Lion G3 Series Hmi Operator Panels
Red Lion Operator Interface With Tft Color Display
Refrigeration Dryers
Replacement Pressure Transducers Sensors For Digi Sense Pressure Meters
Rhodium On Alumina Powder
Ribonuclease A From Bovine Pancreas
Ricca Chemcial Glacial Acetic Acid
Ricca Chemical Acetonitrile
Ricca Chemical Boric Acid
Ricca Chemical Company Sodium Hydroxide Sulfide Solutions For Kjeldahl
Ricca Chemical Company Sodium Hydroxide W V Solutions 72026
Ricca Chemical Company Sodium Hydroxide W W Solutions
Ricca Chemical Company Verispec 19 Element Calibration Standard
Ricca Chemical Company Verispec 22 Element Calibration Standard
Ricca Chemical Company Verispec 30 Element Calibration Standard
Ricca Chemical Company Verispec 4 Element Calibration Standard
Ricca Chemical Company Verispec 5 Element Calibration Standard
Ricca Chemical Company Verispec 6 Element Calibration Standard
Ricca Chemical Company Verispec Arsenic 3 Icp Standard
Ricca Chemical Company Verispec Barium Aas Standard
Ricca Chemical Company Verispec Barium Icp Standard
Ricca Chemical Company Verispec Benzoate Ion Chromatography Standard
Ricca Chemical Company Verispec Bromide Ion Chromatography Standard
Ricca Chemical Company Verispec Cadmium Icp Standard
Ricca Chemical Company Verispec Cerium Icp Standard
Ricca Chemical Company Verispec Cesium Aas Standard
Ricca Chemical Company Verispec Copper Aas Standard
Ricca Chemical Company Verispec Diethanolamine Ion Chromatography Standard
Ricca Chemical Company Verispec Dysprosium Aas Standard
Ricca Chemical Company Verispec Dysprosium Icp Standard
Ricca Chemical Company Verispec Erbium Icp Standard
Ricca Chemical Company Verispec Europium Aas Standard
Ricca Chemical Company Verispec Europium Icp Standard
Ricca Chemical Company Verispec Gadolinium Aas Standard
Ricca Chemical Company Verispec Gadolinium Icp Standard
Ricca Chemical Company Verispec Gallium Aas Standard
Ricca Chemical Company Verispec Hafnium Aas Standard
Ricca Chemical Company Verispec Hafnium Icp Standard
Ricca Chemical Company Verispec Holmium Aas Standard
Ricca Chemical Company Verispec Holmium Icp Standard
Ricca Chemical Company Verispec Hydrogen Phthalate Ion Chromatography Standard
Ricca Chemical Company Verispec Indium Aas Standard
Ricca Chemical Company Verispec Lactate Ion Chromatography Standard
Ricca Chemical Company Verispec Lutetium Icp Standard
Ricca Chemical Company Verispec Magnesium Aas Standard
Ricca Chemical Company Verispec Maleate Ion Chromatography Standard
Ricca Chemical Company Verispec Manganese Icp Standard
Ricca Chemical Company Verispec Neodymium Aas Standard
Ricca Chemical Company Verispec Nickel Aas Standard
Ricca Chemical Company Verispec Niobium Aas Standard
Ricca Chemical Company Verispec Nitrite No Ion Chromatography Standard
Ricca Chemical Company Verispec Oxalate Ion Chromatography Standard
Ricca Chemical Company Verispec Palladium Aas Standard
Ricca Chemical Company Verispec Palladium Icp Standard
Ricca Chemical Company Verispec Potassium Aas Standard
Ricca Chemical Company Verispec Rhenium Aas Standard
Ricca Chemical Company Verispec Rhodium Aas Standard
Ricca Chemical Company Verispec Rhodium Icp Standard
Ricca Chemical Company Verispec Rubidium Aas Standard
Ricca Chemical Company Verispec Ruthenium Icp Standard
Ricca Chemical Company Verispec Samarium Aas Standard
Ricca Chemical Company Verispec Samarium Icp Standard
Ricca Chemical Company Verispec Scandium Icp Standard
Ricca Chemical Company Verispec Silicon Aas Standard
Ricca Chemical Company Verispec Silver Aas Standard
Ricca Chemical Company Verispec Silver Icp Standard
Ricca Chemical Company Verispec Specialty Icp Standards
Ricca Chemical Company Verispec Strontium Aas Standard
Ricca Chemical Company Verispec Tantalum Aas Standard
Ricca Chemical Company Verispec Thallium Aas Standard
Ricca Chemical Company Verispec Thiocyanate Ion Chromatography Standard
Ricca Chemical Company Verispec Tin Aas Standard
Ricca Chemical Company Verispec Trimethylamine Ion Chromatography Standard
Ricca Chemical Company Verispec Ytterbium Icp Standard
Ricca Chemical Company Verispec Zirconium Aas Standard
Ricca Chemical Ethylene Glycol
Ricca Chemical Hydrofluoric Acid Solutions
Ricca Chemical Nitric Acid V V Solutions
Ricca Chemical Perchloric Acid Solutions
Ricca Chemical Sulfuric Acid Acs Grade
Ricca Chemical Sulfuric Acid W W Solutions
Roller Pumps
Rosolic Acid
Rubidium Standard
Ruthenium
Ruthenium On Carbon
Saccharin
Saccharin Sodium
Safety Eyewear Holders
Safranine Aqueous Alcoholic Counterstain For Gram Staining
Sampling Tubes 5437
Sarcosine
Sartorius Anti Theft Devices
Sartorius Cubis Mse Simi Micro Balances With High Contrast Lcd
Sartorius Interface Cables
Sartorius Output Relay Kits
Scienceware Spinring Stirbars Ptfe
Scienceware Switch Grid Tube Racks
Sealless Pvdf Self Priming Centrifugal Pumps
Self Cleaning Industrial Ph Electrodes With In Line Assemblies
Set Point Modules For Cole Parmer Gas And Liquid Flow Controllers
Shimpo Force Gauge Test Stands
Shock Data Loggers
Signet Temperature Sensors
Silicon Plasma Grade Reference Standard
Silver Atomic Absorption Standard Solution
Silver I Fluoride
Silver Nitrate
Silver Standard
Silver Sulfate
S N Fmoc 3 Thienylalanine
Sodium
Sodium Acetate 0 200 Molar M 5
Sodium Acetate Anhydrous
Sodium Borodeuteride
Sodium Caprylate
Sodium Chloride Ndash Tris Ndash Edta Ste Buffer 10X 26256
Sodium Deuteroxide
Sodium Formaldehydesulfoxylate Dihydrate
Sodium Lauryl Sulfate
Sodium Meta Silicate Nonahydrate
Sodium Meta Silicate Pentahydrate
Sodium Nitrite 23478
Sodium Nitrite 5 W W Aqueous Solution
Sodium Oxalate 0 100 Molar M 10
Sodium Phosphate Dibasic Anhydrous 23480
Sodium Phosphate Monobasic Dihydrate
Sodium Phosphate Tribasic Dodecahydrate Reagent Acs
Sodium Sulfanilate Hydrate
Sodium Sulfide Nonahydrate
Sodium Tartrate 10 W W Aqueous Solution
Sodium Tetrafluoroborate
Sodium Thiocyanate 0 100 Normal N 10
Sodium Triacetoxyborohydride 38176
Solenoid Operated Pinch Valves For Metric Tubing
Solvent Recovery Head
Spc Oil Sorbents
Spectrum Chemical 1 10 Phenanthroline Monohydrochloride Monohydrate
Spectrum Chemical 1 1 1 3 3 3 Hexafluoro 2 Propanol
Spectrum Chemical 1 1 2 2 Tetrachloroethane
Spectrum Chemical 1 2 3 4 Tetrahydronaphthalene
Spectrum Chemical 1 2 Cyclohexylenedinitrilo Tetraacetic Acid
Spectrum Chemical 1 3 Dimethoxybenzene
Spectrum Chemical 1 3 Dimethylaminopropyl 3 Ethylcarbodiimide Hcl
Spectrum Chemical 1 3 Dioxolane
Spectrum Chemical 1 3 Propanediol
Spectrum Chemical 1 5 Diphenylcarbohydrazide
Spectrum Chemical 1 6 Hexanediamine
Spectrum Chemical 1 6 Hexanediamine 70 Solution In Water
Spectrum Chemical 1 6 Hexanediol
Spectrum Chemical 1 Bromobutane
Spectrum Chemical 1 Bromonaphthalene
Spectrum Chemical 1 Bromooctane
Spectrum Chemical 1 Bromopropane
Spectrum Chemical 1 Decanesulfonic Acid Sodium Salt
Spectrum Chemical 1 Dodecane Sulfonic Acid Sodium Salt
Spectrum Chemical 1 Heptanol
Spectrum Chemical 1 Methylnaphthalene
Spectrum Chemical 1 Nitroso 2 Naphthol
Spectrum Chemical 1 Pentanesulfonic Acid Sodium Salt
Spectrum Chemical 1 Pentanol
Spectrum Chemical 1 Phenyl 3 Methyl 5 Pyrazolone
Spectrum Chemical 2 2 2 Trifluoroethanol
Spectrum Chemical 2 2 4 Trimethylpentane
Spectrum Chemical 2 2 6 6 Tetramethylpiperidinyloxy Free Radical
Spectrum Chemical 2 2 Butoxyethoxy Ethyl Acetate
Spectrum Chemical 2 2 Dimethoxypropane
Spectrum Chemical 2 4 Diaminophenol Dihydrochloride
Spectrum Chemical 2 4 Pentanedione
Spectrum Chemical 2 6 Dichloroquinone 4 Chloroimide
Spectrum Chemical 2 6 Dimethoxyphenol
Spectrum Chemical 2 6 Toluene Diisocyanate
Spectrum Chemical 2 7 Dihydroxynaphthalene
Spectrum Chemical 2 Acetonaphthone
Spectrum Chemical 2 Acrylamido 2 Methyl Propanesulfonic Acid
Spectrum Chemical 2 Butoxyethanol
Spectrum Chemical 2 Butyne 1 4 Diol
Spectrum Chemical 2 Chlorobenzoic Acid
Spectrum Chemical 2 Deoxy D Glucose
Spectrum Chemical 2 Ethoxybenzoic Acid
Spectrum Chemical 2 Ethyl 2 Hydroxymethyl 1 3 Propanediol
Spectrum Chemical 2 Ethylhexanoic Acid
Spectrum Chemical 2 Iodopropane
Spectrum Chemical 2 Mercaptobenzothiazole
Spectrum Chemical 2 Mercaptoethanol
Spectrum Chemical 2 Methyl 1 Butanol
Spectrum Chemical 2 Naphthol
Spectrum Chemical 2 Nitrobenzaldehyde
Spectrum Chemical 2 Nitropropane
Spectrum Chemical 2 Nonanol
Spectrum Chemical 2 Pentanol
Spectrum Chemical 2 Phenylphenol
Spectrum Chemical 2 Pyrrolidone
Spectrum Chemical 2 Thiophenecarboxylic Acid
Spectrum Chemical 3 3 Thiodipropionic Acid
Spectrum Chemical 3 5 Dinitrosalicylic Acid
Spectrum Chemical 3 Anilino 2 3 4 5 Trimethoxybenzyl Acrylonitrile
Spectrum Chemical 3 Bromophenol
Spectrum Chemical 3 Bromopropene
Spectrum Chemical 3 Chloroperoxybenzoic Acid
Spectrum Chemical 3 Heptanone
Spectrum Chemical 3 Methylbutyraldehyde
Spectrum Chemical 3 Nitrobenzaldehyde
Spectrum Chemical 3 Trimethoxysilyl Propyl Methacrylate
Spectrum Chemical 4 Amino 3 Hydroxy 1 Naphthalene Sulfonic Acid
Spectrum Chemical 4 Aminopyridine
Spectrum Chemical 4 Bromoacetanilide
Spectrum Chemical 4 Bromoacetophenone
Spectrum Chemical 4 Chlorobenzaldehyde
Spectrum Chemical 4 Chlorobenzoic Acid
Spectrum Chemical 4 Dimethylaminopyridine
Spectrum Chemical 4 Hydroxyisophthalic Acid
Spectrum Chemical 4 Methoxyphenol
Spectrum Chemical 4 Methyl 2 Pentanol
Spectrum Chemical 4 Methylamino Phenol Sulfate
Spectrum Chemical 4 Methylmorpholine N Oxide
Spectrum Chemical 4 Nitroaniline Powder
Spectrum Chemical 4 Toluenesulfonic Acid Pyridine Salt
Spectrum Chemical 4 Vinylpyridine
Spectrum Chemical 5 Aminosalicylic Acid
Spectrum Chemical 7 Hydroxy 4 Methylcoumarin
Spectrum Chemical 7 Hydroxycoumarin
Spectrum Chemical 8 Anilino 1 Naphthalenesulfonic Acid
Spectrum Chemical 8 Hydroxy 1 3 6 Pyrenetrisulfonic Acid Trisodium Salt
Spectrum Chemical Acenaphthene
Spectrum Chemical Aces Biological Buffer
Spectrum Chemical Acetamide
Spectrum Chemical Acetate Buffer Ts U S P Test Solution
Spectrum Chemical Acetophenone
Spectrum Chemical Acetyl Chloride
Spectrum Chemical Acid Fuchsin
Spectrum Chemical Acrylamide
Spectrum Chemical Adenine Synthetic
Spectrum Chemical Adipyl Chloride Reagent
Spectrum Chemical Agar
Spectrum Chemical Alanine
Spectrum Chemical Allylamine
Spectrum Chemical Alpha Tocopherol
Spectrum Chemical Aluminum Acetate Dibasic
Spectrum Chemical Aluminum Ammonium Sulfate Dodecahydrate
Spectrum Chemical Aluminum Atomic Absorption Standard
Spectrum Chemical Aluminum Chloride
Spectrum Chemical Aluminum Fluoride
Spectrum Chemical Aluminum Hydroxide
Spectrum Chemical Aluminum Metal
Spectrum Chemical Aluminum Nickel Catalyst Raney Type Alloy
Spectrum Chemical Aluminum Sodium Sulfate
Spectrum Chemical Aluminum Stearate
Spectrum Chemical Ammonium Acetate Buffer Solutions
Spectrum Chemical Ammonium Acetate Solutions
Spectrum Chemical Ammonium Bicarbonate
Spectrum Chemical Ammonium Bromide
Spectrum Chemical Ammonium Chloride Ammonium Hydroxide
Spectrum Chemical Ammonium Chloride Solutions
Spectrum Chemical Ammonium Fluoride
Spectrum Chemical Ammonium Metavanadate
Spectrum Chemical Ammonium Phosphate
Spectrum Chemical Ammonium Phosphate Dibasic
Spectrum Chemical Ammonium Phosphate Dibasic Solutions
Spectrum Chemical Ammonium Phosphate Monobasic
Spectrum Chemical Ammonium Sulfate
Spectrum Chemical Ammonium Sulfate Solutions
Spectrum Chemical Ammonium Tartrate
Spectrum Chemical Anhydrous Dibasic Calcium Phosphate
Spectrum Chemical Aniline Blue Water Soluble
Spectrum Chemical Anthraquinone
Spectrum Chemical Anthrone
Spectrum Chemical Antimony Atomic Absorption Standard
Spectrum Chemical Antimony Metal
Spectrum Chemical Antimony Pentachloride
Spectrum Chemical Antimony Trichloride
Spectrum Chemical Antimony Trisulfide
Spectrum Chemical Arsenazo Iii
Spectrum Chemical Arsenic Plasma Emission Standard
Spectrum Chemical Arsenic Trioxide
Spectrum Chemical Aurin Tricarboxylic Acid
Spectrum Chemical Azomethine H
Spectrum Chemical Azure Ii Eosinate
Spectrum Chemical Barbituric Acid
Spectrum Chemical Barium Diphenylaminesulfonate Solutions
Spectrum Chemical Barium Nitrate
Spectrum Chemical Barium Perchlorate Anhydrous
Spectrum Chemical Barium Sulfide
Spectrum Chemical Bathocuproine
Spectrum Chemical Bathophenanthroline
Spectrum Chemical B Carotene
Spectrum Chemical Benedict S Reagent
Spectrum Chemical Benzamidine Hydrochloride
Spectrum Chemical Benzimidazole
Spectrum Chemical Beryllium Atomic Absorption Standard
Spectrum Chemical Beryllium Plasma Emission Standard
Spectrum Chemical Beta Tetralone
Spectrum Chemical Biscyclohexanone Oxaldihydrazone
Spectrum Chemical Bismuth Subnitrate
Spectrum Chemical Bismuth Sulfite
Spectrum Chemical Bismuth Trioxide
Spectrum Chemical Bis Trimethylsilyl Acetamide
Spectrum Chemical Biuret Reagent Solution
Spectrum Chemical Boron Trifluoride Etherate
Spectrum Chemical Brilliant Green Solutions
Spectrum Chemical Bromocresol Green Solutions