About Piston Pumps
The video illustrates the advantages Masterflex® Ismatec® piston pump drive with MasterflexLive® and the Rh Series piston pump heads can bring to your fluid process such as meeting high pressure requirements or precisely pumping viscous fluids. The fluid chamber is precisely machined to create a specific volume that is pumped at every revolution. It is perfect for dosing or fluid metering applications such as pharmaceutical and chemical downstream processing or the lab.