Lab Equipment Demo: Cole-Parmer Freezer/Mill® Cryogenic Grinders in Action
Watch the Cole-Parmer Freezer/Mill® Cryogenic Grinders in Action
Description
Learn more about and this Freezer/Mill® collection and watch a demo. These programmable cryogenic grinders are specifically designed for milling and grinding tough or temperature-sensitive samples. Get to know more about the 6775 small Freezer/Mill® that accomodates samples up to 5 g, the 6875 large Freezer/Mill® can accommodate samples up to 100 g. And, the largest Freezer/Mill® 6875D is a dual-grinding mill allows up to 200 g per sample, 100 g per chamber.