Flow Meter Selection Guide
Choose The Best Flow Meter For Your Application
When selecting a flow meter consideration must be given to:
  • Flow measurement type - momentum (velocity), volumetric or mass flow measurement
  • Media - type of media (liquid, gas or slurry) and any special condition such as particulates in the media and viscosity of the media
  • Media conditions - pressure and temperature of media and whether media conditions are likely to remain constant or vary
  • Flow range - required flow range of media (min and max readings required)
  • Accuracy - required accuracy of the readings
  • Environmental considerations - special installation considerations such as hygienic installation, installing in to an ATEX zone or requiring tamper proof readings
For special or unique flow control applications use the helpful flow meter application table below to narrow your selection.
Flow Meter Selection Table by Application
Table scrolls horizontally
Application Parameter Gases Liquids Flow Meter Type
Low-flow measurement/control (some types as low as 1 sccm air and 0.1 mL/min water, max of range)
Gas mass
Coriolis
Differential pressure
Indicators/switches
Turbine
Variable area
Pelton wheel
Vortex
Gear
Paddle wheel
Magnetic
Ultrasonic
High-flow measurement/control (some types as high as 2300 scfm gas and 2600 GPM liquid)
Gas mass
Coriolis
Differential pressure
Indicators/switches
Variable area
Pelton wheel
Vortex
Paddle wheel
Magnetic
Turbine
Ultrasonic
High pressure
(500 psi and up)
Gas mass
Coriolis
Switches
Variable area
Gear
Turbine
Ultrasonic
High temperature (200°F and up)
Coriolis
Differential pressure
Vortex (including steam)
Variable area
Ultrasonic
Turbine
Magnetic
Paddle wheel
Gear
High-accuracy measurement (error at or below 1%)
Differential pressure
Pelton wheel
Gas mass
Coriolis
Vortex
Gear
Paddle wheel
Magnetic
Turbine
Ultrasonic
Aggressive or pure fluids
Gas mass
Turbine
Switches
Variable area
Gear
Paddle wheel
Magnetic
Ultrasonic
Vortex
Fluids with heavy particulates or slurries
Coriolis
Magnetic
Ultrasonic
High-viscosity fluids
Coriolis
Gear
Magnetic
No wired power available
Differential pressure
Flow Indicators
Turbine
Variable area
Gear
Paddle wheel
Ultrasonic
Intrinsically safe
Variable area
Turbine
Large pipe sizes
(3" or larger)
Coriolis
Ultrasonic
Magnetic
Paddle wheel
Sanitary
(meets 3A standards)
Turbine
Magnetic