Masterflex: How To Load Tubing - EZ Load
Masterflex pumps are designed to make tubing changes quick and easy. To load tubing simply follow the two steps below:
Image loading tubing to a peristaltic pump
  1. Rotate lever to left to open head.
  2. Load tubing.
  3. Close head.
  4. Adjust side tubing retainers to hold tubing.