Masterflex Ismatec Reglo ICC Peristaltic Pump VIdeo
The Masterflex Reglo Independent Channel Control (ICC) pump’s unique feature is the capability to set a different flow rate and direction in each of up to four channel - this means you are getting four pumps in one small package. Using peristaltic technology the pump action is precise and low shear – perfect for perfusion and “recipe” based pumping. The tubing is mounted into the pump cartridge using stops or flags installed onto the tube which ensures a standardized tension in the occlusion bead so you know you are getting the correct flow in each channel. The below video illustrates this unique ability of the Masterflex Reglo ICC pump to precisely control each channel.