Ismatec tubing cassettes: CA and MS/CA types
Ismatec Click 'n' Go™ Cassettes allow for easy and rapid tubing exchange for peristaltic pumps. There are two types of tubing cassettes available:
CA Cassettes
CA-Cassette for 2-stop tubing (CA Tubing)Large CA cassettes are suitable for use with the IPC and IPC-N series, low flow, high accuracy peristaltic pumps. CA cassettes accept 2-stop peristaltic tubing with a 150 mm gap between the stoppers. Suitable tubing sizes range from 0.13 to 2.79 mm ID. Pressure lever equipped CA cassettes allow to set a different tubing pressure for each channel. Depending on the application, tubing material and diameter, an optimally adjusted tubing pressure can be set. To maintain constant flow rates, it may be necessary to periodically adjust the tubing pressure. Using pressure lever equipped cassettes usually increases the maximum achievable differential pressure of a peristaltic pump, but potentially adversely effect pump tubing life.
When ordering CA cassettes, the replacement adapters for CA cassettes are not included and need to be ordered separately.
2-stop tubing (CA Tubing)
CA cassettes are suitable for:
  • 2-stop (CA) peristaltic tubing
  • gap between tubing stoppers of 152 mm
  • Tubing ID's from 0.13 to 3.17 mm
Order Replacements Here
Please Note: the adapters 07618-65 are not included with the cassettes and need to be ordered separately. They are however included with the pump system
MS/CA Cassettes
MS/CA Cassette for 3-stop tubingThe small MS/CA cassettes are suitable for use with the Reglo Digital and ICC series. MS/CA cassettes accept 3-stop peristaltic (MS/CA) tubing with a 72 mm gap between the stoppers. Suitable tubing sizes range from 0.13 to 3.17 mm ID. Pressure lever equipped MS/CA cassettes allow to set a different tubing pressure for each channel. Depending on the application, tubing material and diameter, an optimally adjusted tubing pressure can be set. To maintain constant flow rates, it may be necessary to periodically adjust the tubing pressure. Using pressure lever equipped cassettes usually increases the maximum achievable differential pressure of a peristaltic pump but can adversely affect tubing life. Standard MS/CA cassettes come without the pressure lever. Consistent pressure is applied with a pre-installed, stainless-steel spring.
3-stop tubing (MS/CA Tubing)
MS/CA cassettes are suitable for:
  • 3-stop (MS/CA) peristaltic tubing
  • gap between tubing stoppers of 72 mm
  • Tubing ID's from 0.13 to 3.17 mm
Order Replacements Here