Selecting Your Tubing
To complete your Ismatec pumping system, order two- or three-stop color-coded tubing or nominal tubing. A variety of materials and sizes are available, as well as extension tubing.
Tubing Types
Two-Stop Tubing
Use two-stop tubing in standard-sized cartridges and tubing bed sets. Choose from the seven tubing material formulations below. Extension tubing is also available. Tubing stops are color-coded according to tubing ID for quick identification. Stops hold the tubing at the optimal tension necessary for pump head rollers to occlude the tubing. The result is reliable pumping and repeatable flow rates.
Three-Stop Tubing
Use three-stop tubing in minicartridges. Choose from the seven tubing material formulations below. Extension tubing is also available. Tubing stops, color-coded according to tubing ID, hold the tubing at the optimal tension necessary for pump head rollers to occlude the tubing. The result is reliable pumping and repeatable flow rates. Three stops give you two sections of tubing to use instead of just one. When one section of tubing fatigues, a fresh section is ready to use.
Nominal Tubing
Use nominal tubing with the quick-change and tubing bed pumps. Choose from seven tubing material formulations below.
Tubing Material Information
Table scrolls horizontally
Material | Apperance | Temperature | Life at 100 rpm | Advantages | Application |
Tygon | Clear | -50 to 74°C | 300 hours | Clear and economical | Acids, alkalies, viscous fluids |
Tygon LFL | Clear | -50 to 74°C | 300 hours | Pharmaceutical grade.Longest flex life of all Tygon | Acids, alkalies, viscous fluids, pharmaceuticals |
Silicone (peroxide cured) | Translucent | -60 to 260°C | 800 hours | Excellent biocompatibility | Sterile fluids |
Silicone (platinum cured) | Translucent | -60 to 260°C | 800 hours | Very low leaching of organic compounds | Sterile fluids |
Norprene | Opaque, black | -50 to 130°C | 1400 hours | Long life | Acids, alkalies, general purpose |
Viton | Opaque, black | -20 to 205°C | 160 hours | Resists most chemicals | Acids, alkalies, solvents, viscous fluids |
PharMed | Opaque, beige | -50 to 130°C | 1400 hours | Pharmaceutical grade | Acids, alkalies, sterile fluids, viscous fluids |
How to order 2- and 3- stop tubing
Choose either 2- or 3- stop tubing per your pump’s requirements