Agar Dispensing Pump Application
Masterflex® Pharmaceutical Applications
Agar Dispensing Pump
Dispenses samples of agar from 0.05 to 8000 mL
Highly accurate performance: ±0.5%
No valves to clog
Easily sterilized
EASY-LOAD® II pump heads ensure low pulsation and accurate performance
Silicone tubing is recommended for purity
Pump product shown*
Pump system 77310-00, including:
Pump = 77200-62
Tubing = 96440-15
Drive = 77310-50
Also shown:*
14200-24 (nutrient agar medium)