Batch Caustic Dispensing Pump Application
Masterflex® Industrial Applications
Batch Caustic Dispensing Pump
For batch transfer and corrosive fluid dispensing
Flow rates up to 30 LPM with B/T® 90 tubing
Ideal for viscous or shear-sensitive liquids
Digital display monitors flow rate, dispense volume, copy number and rpm
Remote control capabilities
Modular design for convenient component placement
RAPID-LOAD® pump head makes changing tubing quick and easy
IP56 wash-down controller and drive are protected from water spray
TYGON® LFL tubing—long life, broad chemical resistance, and high dielectric constant
Pump product shown*
Pump = 77111-70
Tubing = 06440-91