Bioprocess Container Pump Application
Masterflex® Pharmaceutical Applications
Bioprocess Container Pump
Accurately pumps nutrient media from a bulk source into Flexboy® containers
Clear TYGON® LFL tubing for long tubing life and easy flow monitoring; can be gamma irradiated
Flow rates: 0.5 to 15 LPM with I/P® 88 tubing
Monitor flow rate, volume, copy number, rpm
Easy calibration—store 1 value per tubing size
IP56 rated—wash down after use
Pump product shown*
Pump = 77600-62
Tubing = 06440-88
Drive = 07592-30