Masterflex B/T Tubing Options
Masterflex B/T® Tubing Options
Achieve top performance from your Masterflex B/T pump heads when you use precision-extruded tubing. Tubing is manufactured to tight tolerances that match our B/T pump heads. Choose from eight different tubing formulations:
  • Pharmed®
  • BioPharm Silicone
  • C-Flex
  • Silicone (platinum-cured)
  • Silicone (peroxide cured)
  • Tygon LFL
  • BioPharm Silicone (platinum-cured)
  • BioPharm Plus Silicone (platinum-cured)
    B/T Precision Tubing Specifications
    Tubing size B/T 86B/T 88B/T 90B/T 92
    Inside diameter (nominal) 0.37" (9.5 mm) 0.5" (12.7 mm) 0.75" (19.0 mm) 1" (25.4 mm)
    Hose barb size (nominal) 0.37" (9.5 mm) 0.5" (12.7 mm) 0.75" (19.0 mm) 1" (25.4 mm)
    Flow range with 12 to 350 rpm drive
    in mL/min. Values in ( ) obtained with High-Performance pump head
    0.3 to 7.5 LPM (0.07 to 2 GPM) 0.5 to 15 LPM (0.15 to 4 GPM) 1 to 30 LPM (0.3 to 8 GPM) 1.5 to 45 LPM (0.4 to 12 GPM)
    Maximum pressure, continuous** 20 psig (1.4 bar)
    Maximum pressure, intermittent** 20 psig (1.4 bar)
    Maximum vacuum** 26" Hg (660 mm Hg)
    Suction lift 29 ft H2O (8.8 m H2O)
    **Actual performance varies depending on tubing material.