Carboy Transfer Application
Masterflex® Research & Development Applications
Carboy Transfer
Transfers chemicals at flow rates up to 2300 mL/min (36 GPH)
EASY-LOAD® pump head for quick changes between processes
Excellent chemical resistance
Easy clean-up
Self-priming, non-siphoning
Reversible motor
Substitute tubing for compatibility with various chemicals
Pump product shown*
Pump = 07518-60
Tubing = 06424-18
Drive = 07553-70