Composite Sampler Pump Application
Masterflex® Environmental & Special Applications
Composite Sampler
Program operation to meet EPA sampling requirements
Dual-line, 24-character, backlit adjustable LCD display with glow in the dark keypad
Locking latches prevent tampering; sampler floats up to 30 minutes if dropped in water
Complete composite sampler system includes Easy-Load® pump head and L/S® tubing.
Passes through 18" diameter manhole
Up to a 90-day standby for a remote signal
Operates on an internal rechargeable battery, 12 VDC, or 115/230 VAC
go BACK TO TOP