Masterflex Connection Newsletter Vol.3-3 - Specialty pump tubing is the winner in those challenging applications
Volume 3, Number 3, 2001
Survivors!
Specialty pump tubing is the winner in those challenging applications
Masterflex® tubing pumps are ideal for pumping chemicals. The peristaltic design eliminates contact between moving parts and the fluid. And for those applications that present a special challenge, requiring just the right chemically resistant tubing, consider these Masterflex® tubing formulations:
  • Tygon Fuel & Lubricant
- Ideal for transporting hydrocarbons, petroleum products, and distillates. Suitable for gasoline, kerosene, heating oils, cutting compounds, and glycol-based coolants. Minimum extractability and low gas permeability. High dielectric constant.
  • Viton & Viton FDA
- Our most chemically resistant tubing; resistant to corrosives, solvents, and oils at elevated temperatures. Low gas permeability. Available in FDA compliant formulation.
  • Tygon 2075
- This ultra chemical-resistant tubing is ideal for use with highly aggressive chemicals—acids, bases, ketones, salts, and alcohols. Plasticizer free, smoother inner surface (inhibits particulate build-up), hydrophobic, minimal outgassing, safe disposal via incineration, and nontoxic. Contact our Application Specialists for information.
  • PTFE
- Excellent chemical resistance. Maintains high purity; will not leach or absorb into solutions. High-pressure capability up to 100 psi. Clear for easy flow monitoring. Very low gas permeability. Smooth bore.
For more information on chemical compatibility, please call our Application Specialists at (800) MASTERFLEX, go to masterflex.com for online compatibility tables, or ask for your FREE Tubing Compatibility Poster.
Tygon Fuel & Lubricant Tubing
Viton Tubing
FDA Viton Tubing
Tygon 2075 Tubing
PTFE Tubing