Filling Station Pump Application
Masterflex® Research & Development Applications
Filling Station Pump
Fastest Masterflex® benchtop dispenser
Ideal for accurate, large volume dispensing
Copy sample volumes: 1.5 mL to 1000 L
Easy to calibrate
Self-priming
Non-siphoning
Excellent chemical resistance
Start pump with footswitch or contact closure
EASY-LOAD® pump head facilitates quick, easy tubing changes
Pump product shown*
Pump = 77602-10
Tubing = 06509-73
Drive = 07594-00