Fuels & Lubricants Pump Application
Masterflex® Industrial Applications
Fuels & Lubricants Pump
Ideal for transporting hydrocarbons, petroleum products, and distillates
Suitable for gasoline, kerosene, heating oils, cutting fluids, and glycol-based coolants
Minimum extractability
Low gas permeability
High dielectric constant
Compact, portable console package
Pump product shown*
Pump = 07024-52
Tubing = 06401-24
Drive = 07569-00