Gradient Pump Application
Masterflex® Research & Development Applications
Gradient Pump
Remote control operation via PC work station
Highly accurate; easy to program and calibrate
Independently program up to 16 pump/mixers to develop gradients
Wide range of flow rates and pump heads
Low-cost TYGON tubing is clear for visually monitoring fluid flow
Easily sterilized
Pump product shown*
Pump = 07035-21
Tubing = 06440-35
Drive = 07550-20
Also shown:*
50008-32 (Servodyne mixer)
07550-64 (RS-232 cable)
07550-66 (satellite cable for daisy-chaining)