Hazardous Duty Fluid Transfer Application
Masterflex® Industrial Applications
Hazardous Duty Fluid Transfer
Air-powered—suitable for areas where electrical power is unsafe or impractical
No electric power necessary
Ideal for hazardous-duty locations
Intrinsically safe when properly grounded for static electricity
Low-maintenance motor
Connect to existing air supply or compressor
Pump product shown*
Pump = 07019-53
Tubing = 06508-82
Drive = 07589-30