High-Purity Manufacturing Pump Application
Masterflex® Environmental & Special Applications
High-Purity Manufacturing
Suitable for aggressive organic solvents
Flow rates up to 65 mL/min (1 GPH) with PTFE tubing
Withstands pressure up to 6.9 bar (100 psi)
PTFE-wetted parts allow contamination-free pumping
Maintains high fluid purity
Limited chemical exposure—fluid contacts only the tubing
Rigid tubing—pump at higher pressures
Improvement over vacuum pump systems
Pump product shown*
Pump system 77912-10, including:
Pump = 77390-00
Tubing = 77390-60
Drive = 07528-20
Also included:*
Two 1/4" NPT(M) pipe adapters (31321-43) , and one tubing grooving tool (31321-49)
Accessories shown:*
06605-54 (extension tubing)
31321-64 (straight union connector)