Ice Cream Pump Application
Masterflex® Food & Beverage Applications
Ice Cream Pump
Handles viscous materials
Non-contaminating
Self-priming (no valves)
Easy clean-up
Delivers up to 8 LPM (2.1 GPM) with I/P® 73 high-purity silicone tubing
EASY-LOAD® pump head facilitates quick tubing changes between samples
Vary occlusion to handle fruit chunks in the mixture
Variable-speed drive features handles for convenient portability
Pump product shown*
Pump = 77601-60
Tubing = 06418-73
Drive = 77410-10