Juice Production Pump Application
Masterflex® Food & Beverage Applications
Juice Production Pump
Non-contaminating—dispenses food products into ready-to-serve containers
Dispenses samples from 1.5 mL to 1000 L
Dispenses 1900 mL (64 oz) in seconds
Highly accurate: ±0.5%
Wide selection of dispensers
NORPRENE® tubing provides good flow consistency and resistance to acids/alkalies
Pump product shown*
Pump = 07019-21
Tubing = 06402-73
Drive = 07592-30