Liquid Chromatography Pump Application
Masterflex® Research & Development Applications
Liquid Chromatography Pump
System shown delivers flow rates up to 41 mL/min for each channel (max 4 channels)
Cartridge pump head facilitates fast tubing changes between samples
Non-contaminating
Easy clean-up
Choice of tubing sizes for various flow rates and chemical compatibility
Adjustable occlusion—for higher pressure and longer tubing life
Pump product shown*
Pump = 07519-10
Tubing = 96410-14
Drive = 07554-80
Also shown:*
07519-65 (L/S® small cartridge)