Achieve top performance from your Masterflex L/S pump heads when you use precision-extruded tubing. Tubing is manufactured to tight tolerances that match our L/S pump heads. Choose from precision tubing or high-performance precision tubing. High-performance precision tubing has a thicker wall that improves pressure generation, suction lift, ability to pump viscous fluids, and tubing life. Choose from over a dozen different formulations.
Precision Tubing Specifications
Tubing size | L/S 13 | L/S 14 | L/S 16 | L/S 25 | L/S 17 | L/S 18 |
Inside diameter* in. (mm) | 0.03 (0.8) | 0.06 (1.6) | 0.12 (3.1) | 0.19 (4.8) | 0.25 (6.4) | 0.31 (7.9) |
Hose barb size* in. (mm) | 1/16 (1.6) | 1/16 (1.6) | 1/8 (3.2) | 3/16 (4.8) | 1/4 (6.4) | 3/8 (9.5) |
Flow range with 6 to 600 rpm drive mL/min | 0.36 to 36 | 1.3 to 130 | 4.8 to 480 | 10 to 1000 | 17 to 1700 | 23 to 2300 |
Maximum pressure, continuous** | 25 psig (1.7 bar) | 20 psig (1.4 bar) | 15 psig (1.0 bar) | 10 psig (0.7 bar) |
Maximum pressure, intermittent** | 40 psig (2.7 bar) | 35 psig (2.4 bar) | 20 psig (1.4 bar) | 15 psig (1.0 bar) |
Maximum vacuum** | 26" Hg (660 mm Hg) | 20" Hg (510 mm Hg) |
Suction lift | 29 ft H2O (8.8 m H2O) | 22 ft H2O (6.7 m H2O) |
High-Performance Precision Tubing Specifications
Tubing size | L/S 15 | L/S 24 | L/S 35 | L/S 36 |
Inside diameter* in. (mm) | 0.19 (4.8) | 0.25 (6.4) | 0.31 (7.9) | 0.38 (9.7) |
Hose barb size* in. (mm) | 3/16 (4.8) | 1/4 (6.4) | 3/8 (9.5) | 3/8 (9.5) |
Flow range with 6 to 600 rpm drive in mL/min. Values in ( ) obtained with High-Performance pump head | 10 to 1000 (11 to 1100) | 17 to 1700 (18 to 1800) | 23 to 2300 (26 to 2600) | 29 to 2900 (34 to 3400) |
Maximum pressure, continuous** | 25 psig (1.7 bar) | 20 psig (1.4 bar) | 15 psig (1.0 bar) |
Maximum pressure, intermittent** | 40 psig (2.7 bar) | 35 psig (2.4 bar) | 20 psig (1.4 bar) |
Maximum vacuum** | 26" Hg (660 mm Hg) | 24" Hg (610 mm Hg) |
Suction lift | 29 ft H2O (8.8 m H2O) | 27 ft H2O (8.3 m H2O) |
*Nominal size.
**Actual performance varies depending on tubing formulation.