Manufacturing Slurry Pump Application
Masterflex® Industrial Applications
Manufacturing Slurry Pump
Recirculates water in a reaction vessel
Flow rates: 0.3 to 45 LPM (0.07 to 12 gpm)
Pumps IPA, acids, and various oils
Ideal for transfer of viscous or shear-sensitive liquids and abrasives
Polycarbonate front plate provides visibility and safety
Rapid-loading pump head
Wide flow-control range
Pump product shown*
Pump = 07585-30
Tubing = 06485-90