Paint Pigment Pump Application
Masterflex® Industrial Applications
Paint Pigment Pump
Quick and easy tubing changes when changing colors
Flow rates up to 1100 mL/min with High-Performance pump head and L/S® 15 tubing
Digital display monitors flow rate, dispense volume, copy number and rpm
Remote control capabilities
Modular design for convenient component placement
IP56 wash-down controller and drive are protected from fluid spray
Pump product shown*
Pump = 77250-62
Tubing = 06424-15
Drive = 77300-30