Food & Beverage Application
Masterflex® Pumps Food & Beverage Applications
Masterflex® peristaltic pumps can handle a wide range of fluids, from hot water and vitamins to thick molasses. The pump heads are designed for easy tubing exchange, and a wide selection of the drives have "wash-down" capabilities. Most importantly for food applications, Masterflex® pump tubing meets USP Class VI, FDA, NSF and 3A requirements.
Applications-Partial list
Automatic drain cleaners
Flavor concentrate
Glass washing system
Ice cream pump
Juice production
Pizza sauce dispensing
Popcorn butter dispensing
Salad dressing pump
Vitamin A & D Injection
FEATURES BENEFITS
High-purityFluid media only touches ID of tubing-USP, FDA, 3A, NSF approvals.
Easy clean-upSimply change the tubing to clean the pump.
Wide range of tubing materialsCompatible with many foods and chemicals.
Handles viscous materials
Transfer and dispense yogurts, ice cream, and food particles
Easily sterilizedSterilize by ethylene oxide, gamma radiation or autoclave.
Adjustable occlusionOver-occlude for priming; reduce for longer life and particulates.
ReversibleOperate in both directions without disconnecting tubing.
Modular designMix and match pump heads, tubing and drives for many applications.
OEM packagesDesign into your lab dispensers or analytical equipment.