Masterflex Industrial Applications
Masterflex® Industrial Applications
For everyday transferring and metering applications in the laboratory or plant, Masterflex® pumps are the answer. They offer predictable service in continuous-duty areas, while solving many tough pumping applications. Use these pumps to handle waste water, suspended solids, harsh chemicals and more up to 45 LPM (12 GPM). The pumps are easy to use and maintain, and the drives are designed to handle the rugged environment of any plant.
Applications-Partial list
Adhesives for cement
Batch caustic dispensing
Carpet sanitizers/cleaners
Caustic detergents
Chemicals for car washers
Chlorine analyzers
Circuit board manufacturing
Coating for coils
Crankcase oil analyzer
Dispensing glue emulsions
Dyes in fabric manufacturing
Etching chemicals for plate processors
Fuels and lubricants
Hazardous duty fluid transfer
High-purity manufacturing
Inks for lithographs
Laundry chemicals
Lubricator for ball bearings
Microfilm developing machine SOil skimmer
Paint pigments
pH control of effluent
Plating chemicals
Polishing slurry pump
Printing inks
Pulp quality monitoring
Silicone wafer rinse
Smoke-generating machines
Tank transfer pumps
FEATURES BENEFITS
Wide range of tubing materials Compatible with many industrial chemicals.
Non-contaminatingFluid media only touches inside of tubing.
Self-primingSuction lifts up to 8.8 m (29 ft).
Runs DryHandles liquids and gas easily without damaging the pump.
Easy clean-upSimply change the tubing to clean the pump.
Reversible Operate in both directions without disconnecting tubing.
Handles viscous materialsTransfer and dispense oils or fluids in suspension.
Modular designMix and match pump heads, tubing and drives for many applications.
OEM packagesDesign into your own industrial metering and transfer equipment.