Pharmaceutical Applications
Masterflex® Pharmaceutical Applications
Masterflex® pumps are designed to be non-contaminating and non-invasive, making them the pumps of choice in the pharmaceutical industry. Easy-Load® pump heads and a wide range of drives fit many applications, from pumping nutrients or pH adjusters for fermentation, to dispensing cosmetics into small containers. The tubing is traceable to USP Class VI and can be easily sterilized.
Applications-Partial list
FEATURES BENEFITS
Wide range of tubing materials Compatible with many laboratory chemicals.
Non-contaminating Fluid media only contacts ID of tubing.
Easy clean-up Just change the tubing to clean the pump
Multichannel capability Use up to 8 channels simultaneously on one drive.
Sanitary connections on tubing Quick tubing connections within sanitary systems.
Handles viscous materials Dispense agars and suspensions.
Runs dry Use as a low flow aspirator or air/vacuum pump.
Modular design Mix and match pump heads, tubing and drives.
OEM packages Design into your lab dispensers or analytical equipment.