Plating Chemical Pump Application
Masterflex® Industrial Applications
Plating Chemical Pump
Suitable for recirculating electrochemical plating materials
Chemical resistant, easy to clean
IP56 (NEMA 4) controller enclosure
Protection from corrosive environment
NORPRENE® tubing is inexpensive, suitable for daily changes
Flow rate up to 13 LPM with I/P® 88 tubing and High-Performance pump head
Pump product shown*
Pump = 77600-62
Tubing = 06404-88
Drive = 07591-10