Printing Ink Pump Application
Masterflex® Industrial Applications
Printing Ink Pump
Designed to circulate printing ink
Handles viscous materials—no valves to clog
Adaptable to special motors
Easy to clean
Adapter/gear reducer and heavy duty motor reduce flow rate for viscous material
Adjustable occlusion—occlusion of the pump head is reduced to limit abrasion
Tubing can be changed easily and quickly for simple clean-up between print runs
Pump product shown*
Pump System 07588-90, including:
Pump = 77601-10
Tubing = 06404-73
Drive = 07588-60 (included with pump head)