Ultrafiltration Pump Application
Masterflex® Pharmaceutical Applications
Ultrafiltration Pump
Transfers fluid through a filtration system at flow rates up to 13 LPM
Maintains purity of raw materials/chemical solutions and sanitary integrity applications
Allows for end product separations/harvesting
Used in the QC/QA analysis labs for buffer and nutrient media filtration
Extended-life tubing ensures minimum particle entrapment; excellent biocompatibility
Pump product shown*
Pump = 77601-10
Tubing = 96440-73
Drive = 07549-32