Water Sampling Pump Application
Masterflex® Environmental & Special Applications
Water Sampling Pump
Accurately collects water samples
Single-turn potentiometer, ON indicator, and three-position power/reversing switch
Latched sampler floats up to 30 minutes if inadvertently dropped in water
Sample depths to up to 8.8 m (29 ft)
Fluid contacts only the tubing
Operates on rechargeable battery, 12 V car battery, or AC outlet.
Pump product shown*
Pump = 07571-02
Tubing = 96410-25