NEMA enclosure type no. | NEMA definition | IEC enclosure class |
1 | General-purpose. Protects against dust, light, and indirect splashing but is not dust-tight; primarily prevents contact with live parts; used indoors and under normal atmospheric conditions. | IP10 |
2 | Drip-tight. Similar to Type 1 but with addition of drip shields; used where condensation may be severe (as in cooling rooms and laundries). | IP11 |
3 and 3S | Weather-resistant. Protects against weather hazards such as rain and sleet; used outdoors on ship docks, in construction work, and in tunnels and subways. | IP54 |
3R | Intended for outdoor use. Provides a degree of protection against falling rain and ice formation. Meets rod entry, rain, external icing, and rust-resistance design tests. | IP14 |
4 and 4X | Watertight (weatherproof). Must exclude at least 65 GPM of water from 1-in. nozzle delivered from a distance not less than 10 ft for 5 min. Used outdoors on ship docks, in dairies, and in breweries. | IP56 |
5 | Dust-tight. Provided with gaskets or equivalent to exclude dust; used in steel mills and cement plants. | IP52 |
6 and 6P | Submersible. Design depends on specified conditions of pressure and time; submersible in water; used in quarries, mines, and manholes. | IP67 |
7 | Hazardous. For indoor use in Class I, Groups A, B, C, and D environments as defined in the NEC. | |
8 | Hazardous. For indoor and outdoor use in locations classified as Class I, Groups A, B, C, and D as defined in the NEC. | |
9 | Hazardous. For indoor and outdoor use in locations classified as Class II, Groups E, F, or G as defined in the NEC. | |
10 | MSHA. Meets the requirements of the Mine Safety and Health Administration, 30 CFR Part 18 (1978). | |
11 | General-purpose. Protects against the corrosive effects of liquids and gases. Meets drip and corrosion-resistance tests. | |
12 and 12K | General-purpose. Intended for indoor use, provides some protection against dust, falling dirt, and dripping noncorrosive liquids. Meets drip, dust, and rust resistance tests. | IP52 |
13 | General-purpose. Primarily used to provide protection against dust, spraying of water, oil, and noncorrosive coolants. Meets oil exclusion and rust resistance design tests. | IP54 |