Oxygen/Dissolved Oxygen
Oxygen/Dissolved Oxygen
Theory    Sensors     Applications    Temperature Compensation    Salinity Correction    Biochemical Oxygen Demand (BOD)    Measurement Tips
Theory
For a given partial pressure of oxygen in air, the concentration of dissolved oxygen (DO) in saturated pure water is fixed at any one temperature. Electro-metric methods of DO measurement are based on this fixed relationship. By measuring the partial pressure of oxygen and correcting for factors such as temperature and salinity, DO meters can determine the concentration of oxygen in samples.
The concentration of dissolved oxygen is usually expressed in milligrams of oxygen per liter of water (mg/L) or parts per million (ppm). Some meters compare calculated oxygen content with observed concentration and report percent saturation (% sat.).
Sensors
Oxygen Sensor Diagram
Oxygen sensors have a thin organic membrane covering a layer of electrolyte and two metal electrodes. Oxygen diffuses through the membrane at a rate proportional to its partial pressure—the greater the oxygen partial pressure, the more oxygen diffuses through the membrane. Oxygen meters measure the current as oxygen is reduced at the cathode and more oxygen diffuses through the membrane. Since the diffusion current is directly proportional to the concentration of dissolved oxygen, the calibrated meter simply converts measured current into concentration units.
Measuring dissolved oxygen with electrodes is more convenient and more accurate than chemical titration. Two types of oxygen sensors are commonly used:
Polarographic Probes require voltage input from the meter to polarize the electrodes. Since the voltage from an external source may take up to 15 minutes to stabilize, polarographic probes usually need to warm up before use to ensure proper polarization of the electrodes.
Galvanic Probes have electrodes made from two different metals that spontaneously polarize to generate the voltage. Since the voltage is spontaneous rather than supplied by an external source, galvanic probes are always operable and do not require the "warm up" time that polarographic probes need for polarization.
Applications
Dissolved oxygen measurements are used to monitor processes where oxygen content affects reaction rates, process efficiency, or environmental conditions:
  • Waste water treatment
  • Wine production
  • Bio-reactions
  • Environmental water testing
Temperature Compensation
Temperature affects both the solubility and diffusion rate of oxygen; therefore, temperature compensation is necessary for standardized DO measurements. All of our oxygen meters have automatic temperature compensation for various temperature ranges.
Salinity Correction
Since the presence of dissolved salts limits the amount of oxygen that can dissolve in water, the relationship between the partial pressure and concentration of oxygen varies with the salinity of the sample. Most meters feature manual salinity correction to compensate for variations in ionic concentration—simply enter the salinity of the sample in parts per thousand (ppt) to ensure standardized DO measurements.
Biochemical Oxygen Demand (BOD)
The BOD test measures the molecular oxygen utilized in the biodegradation of organic material and the oxidation of inorganic material. By measuring the amount of oxygen dissolved in samples at the beginning and end of a specified incubation period, the relative oxygen requirements of wastewaters, effluents, and polluted waters can be determined.
BODt (mg/L) =
D1 - D2
P
where:
BODt = Oxygen uptake during incubation period t
D1 = DO of diluted sample immediately after preparation (mg/L)
D2 = DO of diluted sample after incubation period t (mg/L)
P = decimal volumetric fraction of sample used
Measurement Tips
Follow these suggestions from our Technical Training Department for accurate measurements:
Accurate Measurements
  1. If using a polarographic probe, allow the instrument to warm up for at least 15 to 30 minutes before calibration or measurement to ensure proper polarization of the sensor. This is particularly important after changing the membrane or cleaning the anode and cathode.
  2. Make sure that the membrane is on tight and that no air bubbles are in the electrolyte solution.
  3. Remove any air bubbles on the probe surface. The probe reads an air bubble as an oxygen-saturated sample.
  4. Calibrate at temperatures as close as possible to the sample temperature. Although most systems have automatic temperature compensation (ATC), the ATC element may take up to 10 minutes to equilibrate.
  5. When calibrating in air, always remove any droplets of water from the probe. The evaporation of any condensation can cool the probe surface.
  6. Always stir samples during measurement. Oxygen consumption by the probe can momentarily reduce the number of molecules at the probe surface. Stirring prevents erroneous measurements due to local variations in sample oxygen concentration.
  7. If the time between measurements is short, leave the instrument on to eliminate the need for repolarization and recalibration.
  8. Replace the membrane as needed, typically every two weeks to one month, depending on your application and the condition of the membrane.
  9. Store the probe in a moist atmosphere to prevent evaporation of the electrolyte solution. Use an electrode storage bottle with a moistened cloth or beaker of water for storage.