No more damaged autosampler needles or sample loss
Introducing Kinesis SureStop Vials
Kinesis SureStop® 9mm Short Thread Autosampler Vials SureStop feature facilitates trouble-free autosampler operation and confidence in analytical results.
- SureStop positioning and optimum vial geometry provides trouble-free autosampler operation
- No more over tightening/deforming septa
- Full confidence in results with no sample evaporation
- Full range of caps and septa
KinesisSureStop 9 mm Short Thread Autosampler Vials have been designed with an over-wind barrier, preventing the cap/septa from being overtightened (see Figure 1). This ensures the septa is not deformed, minimising damage of autosampler needles and preventing sample evaporation due to poor vial-septa-cap sealing. The SureStop vials are fully compatible with all Kinesis Short Thread Caps and Septa, offering the full range of cleanliness and septa injection options. Kinesis SureStop Vials are available in clear, clear with label, and filling lines and amber with label and filling lines.
Optimum Vial-Septa-Cap Sealing – minimal analyte/solvent loss
Solvent loss (methanol) at 40°C for 24 hours was investigated, comparing the Kinesis SureStop Vial with two commercially available standard 9mm short thread vial products. As shown in Figure 2, analyzing 50 vials from two commercially available sources, and comparing with the Kinesis SureStop Vial, shows loss of methanol that would potentially impact reported results. The Kinesis SureStop vial showed little or no solvent loss throughout the test period with either optimal or over tightening.
Ordering Information
The following table provides details of the Kinesis SureStop Vials and hort thread caps and septa.
Description | Pack size | Ordering number |
SureStop Short Thread Vial 9mm, 2ml | 1000 | #98702-78 |
SureStop Short Thread Vial 9mm, 2ml, Label | 1000 | 98702-75 |
SureStop Short Thread Vial 9mm 2ml, Amber, Label | 1000 | 98702-71 |
Short Thread Cap, Clear, Red Rubber/PTFE Septa | 1000 | 98703-34 |
Short Thread Cap, Blue, Red Rubber/PTFE Septa | 1000 | 98703-35 |
Short Thread Cap, Blue, Red Rubber/PTFE Septa | 1000 | 98703-36 |
Short Thread Cap, Clear, Silicone/Red PTFE Septa | 1000 | 98703-47 |
Short Thread Cap, Blue, Silicone/Red PTFE Septa w/slit | 1000 | 98703-54 |
Short Thread Cap, Blue, Silicone/Red PTFE Septa, Ultrabond w/slit | 1000 | 98703-16 |
Additional short thread caps and septa are available on request.