Gregg E. Johnson, Sr. Global Product Manager, Masterflex Bioprocessing, USA.
Pulp & paper operations continuously handle abrasive slurries and highly aggressive chemicals – and this creates huge demands on pumping systems. The recovery of back liquor soap, for example, contains particulate matter, is abrasive, hot, and entrained gases can cause seal failure in certain pump designs. Lime slurries used to produce bleach liquor can easily settle out and clog valves. In addition, maintaining precise, consistent, repeatable flow rates often present major challenges for the metering of dyes, sizing agents, retention aids, and other chemicals.
Solution
Masterflex® L/S® Series peristaltic pumps, are impervious to highly abrasive fluids and gentle on shear sensitive materials. Handling tough, viscous slurries are never a problem due to the pump’s high suction lift capabilities. Plus, there are no glands, valves, or seals to become clogged or wear out. These peristaltic pumps can also run dry. And, unlike many pump designs used in pulp & paper processes, these maintain a precise, constant flow rate with excellent repeatability – providing volumetric accuracies of ± 0.5% of liquid volume, or better.
Benefits
Masterflex hose pumps provide pulp and paper operations with extremely versatile liquid chemical feed and fluid handling solutions with the highest reliability, volumetric accuracy, and long-term operation, plus simple, low maintenance. Masterflex L/S peristaltic pump systems are ideal for handling virtually any chemical, slurry, and additive at pulp and paper operations. The only material the fluid touches is the inside of the tubing, and since no other parts of the pump ever contact the solution, only the tubing ever needs to be cleaned or changed, which is a very quick and simple procedure.
Highly versatile liquid feed & fluid handling solutions
Peristaltic pumps are ideal for handling virtually any chemical, slurry, and additive at pumps and paper operations
Masterflex L/S pumps are the ideal solution for efficiently carrying out the demanding bulk transfer and chemical metering applications of pulp & paper manufacturers.
Aggressive materials, such as phosphoric acid, soda ash and sodium nitrate solution, or Kaolin and clays containing latex and abrasive silica, can lead to defective valves and premature pump wear on other pump types. However, these materials remain inside the pump’s hose or tubing at all times and cannot contaminate and damage the pump or meter. The pump’s non-intrusive design provides a straight flow path with no dead flow areas for slurry to agglomerate, harden, and contaminate the process. Flow rates are accurate up to
± 0.5%. These pumps are innovative, cost-effective peristaltic pumping systems which exhibit low shear force, so it’s gentle on polymers and shear-sensitive slurries, and its variable speed operation allows for precise control of flow or pressure over a wide operating range to maintain process integrity.
For metering process sensitive materials, such as dyes, optical brightening agents, and retention aids, Masterflex pumps provide high accuracy, repeatability, and reliability, especially when compared to diaphragm metering pumps, which often cause excessive downtime for cleaning, stripping and repair.
A well-balanced range of capacities and flexibility combines with excellent repeatability and optimal chemical resistance to provide for precision, high reliability and long-term operation for handling virtually all chemical products for pulp & paper production.
Handles abrasives, shear sensitive materials
- With adjustable 650:1 resolution, bidirectional flow and self-priming capabilities, Masterflex pumps provide for smooth, seamless flow
- Superior performance in viscous and abrasive fluid handling applications such as black liquor soap and lime slurries
- Gentle peristaltic method of pumping prevents surging and will not damage Latex or shear sensitive polymers
- Masterflex L/S pumps are extremely flexible with slurry viscosities
- Masterflex pumps produce very little heat, compared to gear pumps, protecting the integrity of temperature-sensitive dyes and chemical additions
- Peristaltic design means no valves, glands or seals to clean or wear out
- Requires very little maintenance to keep in peak operating condition
- Pumps have an accuracy of ± 0.5 or better and flow rates from a few fluid ounces to multiple gallons per minute
- Pump acts as its own check valve. When the pump stops, the tube stays shut, with no siphoning or dribbling!
- Maintenance-free brushless motor drive (rated for continuous duty) and roller and ball bearing pump head construction produce a robust, powerful pump ideal for long-term operation
- Self-priming for instant startup. And high suction lift provides for operating flexibility. The pump will also operate dry without damage!
Pulp & paper applications
- Dyes – Retention Aids
- Brightening Agents
- Alum – Sizing Agents
- Pulp Chemicals – Latex
- Soda Ash – Phosphoric Acid
- Sodium Nitrate Solution
- Alkalis – Acids – Polymer
- Defoaming Agents
- Titanium Dioxide – Glues
- Clay Solutions – Bleach Liquor
- Lime Slurry – Black Liquor Soap
Masterflex B/T Bulk Transfer Pump System
Four-digit LED display shows flow rate (0.1, 1, or 10 LPM resolution autoranging with tubing size,
bi-directional flow and self-priming
capabilities, user-friendly Masterflex pumps provide precise, easy bulk fluid transfer and dispense