Viscosity and Specific Gravity Values
Viscosity Values and Specific Gravity
Centipoise*
(cp)
Centistokes
(cSt)
Saybolt Second
Universal (SSU)
Typical liquid Specific Gravity
1 1 31 Water 1.0
3.2 4 40 Milk -
12.6 15.7 80 No. 4 fuel oil 0.82-0.95
16.5 20.6 100 Cream -
34.6 43.2 200 Vegetable oil 0.912-0.924
88 110 500 SAE 10 oil 0.88-0.935
176 220 1000 Tomato juice -
352 440 2000 SAE 30 oil0.88-0.935
880 1100 5000 Glycerine 1.26
1561 1735 8000 SAE 50 oil0.88-0.935
1760 2200 10,000 Honey -
3000 4500 20,000 Glue -
5000 6250 28,000 Mayonnaise -
8640 10,800 50,000 Molasses B 1.40-1.49
15,200 19,000 86,000 Sour cream -
17,640 19,600 90,000 SAE 70 oil0.88-0.935
- - - Ink-Printers1.0-1.38
- - - Sulfuric Acid1.83
*Centipoise = centistokes x specific gravity where specific gravity is assumed to be 0.8 (except for water). To find the exact cp of your fluid:
cp = cSt x (weight per gallon x 0.120).