FEATURES | BENEFITS |
Wide range of tubing materials | Compatible with many laboratory chemicals.
|
Non-contaminating | Fluid media only contacts ID of tubing.
|
Easy clean-up | Just change the tubing to clean the pump
|
Multichannel capability | Use up to 8 channels simultaneously on one drive.
|
Sanitary connections on tubing | Quick tubing connections within sanitary systems.
|
Handles viscous materials | Dispense agars and suspensions.
|
Runs dry | Use as a low flow aspirator or air/vacuum pump.
|
Modular design | Mix and match pump heads, tubing and drives.
|
OEM packages | Design into your lab dispensers or analytical equipment.
|